Learn More
Biomatik Corporation Mouse G6PC (Glucose-6-Phosphatase, Catalytic) ELISA Kit
Supplier:  Biomatik Corporation EKF60475
Encompass_Preferred
Mouse G6PC (Glucose-6-Phosphatase, Catalytic) ELISA Kit for detecting G6PC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35576. Target Synonyms: G6PC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6505