Learn More
Biomatik Corporation Human TGFBI/βIG-H3 (Transforming growth factor-beta-induced protein ig-h3) ELISA Kit
Supplier:  Biomatik Corporation EKF60482
Encompass_Preferred
Human TGFBI/βIG-H3 (Transforming growth factor-beta-induced protein ig-h3) ELISA Kit for detecting TGFBI; βIG-H3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15582. Target Synonyms: TGFBI; βIG-H3; Transforming growth factor-beta-induced protein ig-h3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6511