Learn More
Biomatik Corporation Human APOE (Apolipoprotein E) ELISA Kit
Supplier:  Biomatik Corporation EKF60491
Encompass_Preferred
Human APOE (Apolipoprotein E) ELISA Kit for detecting APOE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02649. Target Synonyms: APOE; Apolipoprotein E; Apo-E; MGC1571; apolipoprotein E3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6518