Learn More
Biomatik Corporation Human PAF (PCNA-associated factor) ELISA Kit
Supplier:  Biomatik Corporation EKF60503
Encompass_Preferred
Human PAF (PCNA-associated factor) ELISA Kit for detecting PAF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15004. Target Synonyms: PAF; KIAA0101; PCNA-associated factor; L5; NS5ATP9; PAF15; HCV NS5A-transactivated protein 9; OEATC-1; PCNA-associated factor of 15 kDa; Hepatitis C virus NS5A-transactivated protein 9; Overexpressed in anaplastic thyroid carcinoma 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6529