Learn More
Biomatik Corporation Human MMP-10 (Matrix Metalloproteinase 10) ELISA Kit
Supplier:  Biomatik Corporation EKF60508
Encompass_Preferred
Human MMP-10 (Matrix Metalloproteinase 10) ELISA Kit for detecting MMP-10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09238. Target Synonyms: MMP-10; MMP10; SL-2; STMY2; Stromelysin-2; Transin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6534