Learn More
Biomatik Corporation Mouse Il-11 (Interleukin-11) ELISA Kit
Supplier:  Biomatik Corporation EKF60520
Encompass_Preferred
Mouse Il-11 (Interleukin-11) ELISA Kit for detecting Il-11 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P47873. Target Synonyms: Il-11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6546