Learn More
Biomatik Corporation Mouse AGER (Advanced Glycosylation End Product Specific Receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF60527
Encompass_Preferred
Mouse AGER (Advanced Glycosylation End Product Specific Receptor) ELISA Kit for detecting AGER in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q62151. Target Synonyms: RAGE; AGER. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6552