Learn More
Biomatik Corporation Mouse CXCL9/MIG (Monokine induced by interferon-gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF60531
Encompass_Preferred
Mouse CXCL9/MIG (Monokine induced by interferon-gamma) ELISA Kit for detecting CXCL9; MIG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18340. Target Synonyms: CXCL9; Miγ; MIG; CMK; crg-10; C-X-C motif chemokine 9; Gamma-interferon-induced monokine; Humig; HuMIG; MIGSmall-inducible cytokine B9; monokine induced by gamma interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6555