Learn More
Biomatik Corporation Mouse HSP-60 (Heat Shock Protein 60) ELISA Kit
Supplier:  Biomatik Corporation EKF60540
Encompass_Preferred
Mouse HSP-60 (Heat Shock Protein 60) ELISA Kit for detecting HSP-60 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P63038. Target Synonyms: HSP-60; HSPD1; Heat Shock Protein 60; cpn60; GROEL; SPG13; 60 kDa chaperonin; Chaperonin 60; heat shock 60kD protein 1; chaperonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6563