Learn More
Biomatik Corporation Mouse IL-9 (Interleukin 9) ELISA Kit
Supplier:  Biomatik Corporation EKF60548
Encompass_Preferred
Mouse IL-9 (Interleukin 9) ELISA Kit for detecting IL-9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15247. Target Synonyms: IL-9; Interleukin 9; IL9; HP40; P40; TCGFIII; TCGF3; P40 T-cell and Mast Cell Growth Factor; P40 Cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6571