Learn More
Biomatik Corporation Mouse REN (Renin) ELISA Kit
Supplier:  Biomatik Corporation EKF60550
Encompass_Preferred
Mouse REN (Renin) ELISA Kit for detecting REN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06281. Target Synonyms: REN; Renin; Angiotensinogenase; angiotensin-forming enzyme; HNFJ2; renin precursor; renal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6573