Learn More
Biomatik Corporation Mouse Hif1α (Hypoxia-inducible factor 1-alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60565
Encompass_Preferred
Mouse Hif1α (Hypoxia-inducible factor 1-alpha) ELISA Kit for detecting Hif1α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61221. Target Synonyms: Hif1α; HIF-1α; HIF1A; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; BHLHE78; Class E basic helix-loop-helix protein 78; HIF-1-alpha; HIF1-alpha; hypoxia-inducible factor 1-alpha; Member of PAS protein 1; member of PAS superfamily 1; PAS domain-containing protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6588