Learn More
Biomatik Corporation Human FAS (Tumor necrosisfactor receptor superfamily member 6) ELISA Kit
Supplier:  Biomatik Corporation EKF60574
Encompass_Preferred
Human FAS (Tumor necrosisfactor receptor superfamily member 6) ELISA Kit for detecting FAS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25445. Target Synonyms: FAS; CD95; FASR; TNFRSF6; APO1; APT1; FAS1; FASTM; ALPS1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6597