Learn More
Biomatik Corporation Human APAF1 (Apoptosis Protease Activating Factor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60576
Encompass_Preferred
Human APAF1 (Apoptosis Protease Activating Factor 1) ELISA Kit for detecting APAF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14727. Target Synonyms: APAF1; APAF-1apoptotic protease activating factor 1; apoptotic peptidase activating factor; apoptotic peptidase activating factor 1; apoptotic protease activating factor; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; KIAA0413. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6599