Learn More
Biomatik Corporation Human SERPINC1 (Antithrombin-III) ELISA Kit
Supplier:  Biomatik Corporation EKF60583
Encompass_Preferred
Human SERPINC1 (Antithrombin-III) ELISA Kit for detecting SERPINC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 3.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01008. Target Synonyms: SERPINC1; Antithrombin-III; ATIII; AT3; antithrombin-III; MGC22579; serine; or cysteine proteinase inhibitor; clade C; antithrombin; member 1; serine-cysteine proteinase inhibitor clade C member 1; Serpin C1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6606