Learn More
Biomatik Corporation Rat NAGase (N-Acetyl Beta-D-Glucosaminidase) ELISA Kit
Supplier:  Biomatik Corporation EKF60606
Encompass_Preferred
Rat NAGase (N-Acetyl Beta-D-Glucosaminidase) ELISA Kit for detecting NAGase in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8VIJ5. Target Synonyms: NAGase; N-Acetylhexosaminidase; beta-D-N-acetylhexosaminidase; beta-N-acetyl-D-hexosaminidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6629