Learn More
Biomatik Corporation Human PDGF-AB (Platelet-Derived Growth Factor AB) ELISA Kit
Supplier:  Biomatik Corporation EKF60610
Encompass_Preferred
Human PDGF-AB (Platelet-Derived Growth Factor AB) ELISA Kit for detecting PDGF-AB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04085; P01127. Target Synonyms: Platelet-Derived Growth Factor AB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6633