Learn More
Biomatik Corporation Mouse Prok2 (Prokineticin-2) ELISA Kit
Supplier:  Biomatik Corporation EKF60611
Encompass_Preferred
Mouse Prok2 (Prokineticin-2) ELISA Kit for detecting Prok2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QXU7. Target Synonyms: Prok2; Prokineticin-2; PROK2; BV8; Protein Bv8 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6634