Learn More
Biomatik Corporation Human GLUT2 (Glucose Transporter 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60612
Encompass_Preferred
Human GLUT2 (Glucose Transporter 2) ELISA Kit for detecting GLUT2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11168. Target Synonyms: GLUT2; SLC2A2; GLUT-2; GLUT2 Glucose transporter type 2; liver; SLC2A2; solute carrier family 2; facilitated glucose transporter; member 2; solute carrier family 2; facilitated glucose transporter member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6635