Learn More
Biomatik Corporation Rat Pde5a (cGMP-specific 3',5'-cyclic phosphodiesterase) ELISA Kit
Supplier:  Biomatik Corporation EKF60619
Encompass_Preferred
Rat Pde5a (cGMP-specific 3',5'-cyclic phosphodiesterase) ELISA Kit for detecting Pde5a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O54735. Target Synonyms: Pde5a; cGMP-binding cGMP-specific phosphodiesterase; CGB-PDE; PDE5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6641