Learn More
Biomatik Corporation Rat CTNNb (Catenin, Beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60620
Encompass_Preferred
Rat CTNNb (Catenin, Beta) ELISA Kit for detecting CTNNb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9WU82. Target Synonyms: CTNNB1; CTNNβ1; beta-catenin; catenin; cadherin-associated protein; beta 1; 88kDa; catenin beta-1; CTNNB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6642