Learn More
Biomatik Corporation Human HTRA1 (Serine protease HTRA1) ELISA Kit
Supplier:  Biomatik Corporation EKF60633
Encompass_Preferred
Human HTRA1 (Serine protease HTRA1) ELISA Kit for detecting HTRA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92743. Target Synonyms: HTRA1; Serine protease HTRA1; IGFBP5-protease; PRSS11; ARMD7; High-tempeRature requirement A serine peptidase 1; HtrA serine peptidase 1; L56; ORF480; Serine protease 11; serine; 11; IGF binding; CARASIL; HtrA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6654