Learn More
Biomatik Corporation Human TGFBR1 (TGF-beta receptor type-1) ELISA Kit
Supplier:  Biomatik Corporation EKF60634
Encompass_Preferred
Human TGFBR1 (TGF-beta receptor type-1) ELISA Kit for detecting TGFBR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P36897. Target Synonyms: TGFBR1; AAT5; ALK5; MSSE; SKR4; ALK-5; LDS1A; LDS2A; TGFR-1; ACVRLK4; Transforming growth factor-beta receptor type I; TGF-beta receptor type I; TbetaR-I; TGF-beta type I receptor; Serine; threonine-protein kinase receptor R4; SKR4; Activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ALK-5; ALK5; Activin receptor-like kinase 5; ACVRLK4; ALK-5ALK5; LDS1A; LDS2A; Serine; threonine-protein kinase receptor R4; SKR4; tbetaR-I; TbetaR-I Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6655