Learn More
Biomatik Corporation Human PTPRN (Protein Tyrosine Phosphatase Receptor Type N) ELISA Kit
Supplier:  Biomatik Corporation EKF60642
Encompass_Preferred
Human PTPRN (Protein Tyrosine Phosphatase Receptor Type N) ELISA Kit for detecting PTPRN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q16849. Target Synonyms: PTPRN; IA2; ICA512; PTPRN; R-PTP-N; CA 512; IA2; IA-2; IA-2; PTP; ICA 512; ICA3; ICA512; islet cell antigen 2; Islet cell antigen 512; Islet cell autoantigen 3; protein tyrosine phosphatase; receptor type; N; protein tyrosine phosphatase-like N; PTP IA-2; receptor-type tyrosine-protein phosphatase-like N; R-PTP-N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6661