Learn More
Biomatik Corporation Mouse Fasn (Fatty acid synthase) ELISA Kit
Supplier:  Biomatik Corporation EKF60649
Encompass_Preferred
Mouse Fasn (Fatty acid synthase) ELISA Kit for detecting Fasn in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19096. Target Synonyms: Fasn; Fatty acid synthase; FAS; OA-519; SDR27X1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6666