Learn More
Biomatik Corporation Human ANGPTL4 (Angiopoietin-related protein 4) ELISA Kit
Supplier:  Biomatik Corporation EKF60661
Encompass_Preferred
Human ANGPTL4 (Angiopoietin-related protein 4) ELISA Kit for detecting ANGPTL4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9BY76. Target Synonyms: ANGPTL4; Angiopoietin Like Protein 4; ARP4; FIAF; HFARP; NL2; PGAR; pp1158. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6677