Learn More
Biomatik Corporation Human IGFBP-4 (Insulin-like growth factor-binding protein 4) ELISA Kit
Supplier:  Biomatik Corporation EKF60667
Encompass_Preferred
Human IGFBP-4 (Insulin-like growth factor-binding protein 4) ELISA Kit for detecting IGFBP-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22692. Target Synonyms: IGFBP-4; IGFBP4; BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6682