Learn More
Biomatik Corporation Human NOTCH1 (Neurogenic locus notch homolog protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60681
Encompass_Preferred
Human NOTCH1 (Neurogenic locus notch homolog protein 1) ELISA Kit for detecting NOTCH1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P46531. Target Synonyms: NOTCH1; hN1; Neurogenic locus notch homolog protein 1; TAN1; Translocation-associated notch protein TAN-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6693