Learn More
Biomatik Corporation Human GPC3 (Glypican-3) ELISA Kit
Supplier:  Biomatik Corporation EKF60683
Encompass_Preferred
Human GPC3 (Glypican-3) ELISA Kit for detecting GPC3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51654. Target Synonyms: GPC3; Glypican 3; DGSX; glypican proteoglycan 3; Glypican-3; GTR2-2; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; OCI5; OCI-5; secreted glypican-3; SGB; SGBS; SGBS1SDYS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6695