Learn More
Biomatik Corporation Mouse CXCL1/GROα (Growth Regulated Oncogene Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60687
Encompass_Preferred
Mouse CXCL1/GROα (Growth Regulated Oncogene Alpha) ELISA Kit for detecting CXCL1; GROα in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12850. Target Synonyms: CXCL1; GROα; Growth Regulated Oncogene Alpha; NAP3; GRO1; GRO-A; MGSA; MGSA-A; SCYB1; FSP; CINC-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6699