Learn More
Biomatik Corporation Chicken LHRH/GnRH (Luteinizing Hormone Releasing Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF60702
Encompass_Preferred
Chicken LHRH/GnRH (Luteinizing Hormone Releasing Hormone) ELISA Kit for detecting LHRH; GnRH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P37042. Target Synonyms: GNRH; GnRH associated peptide I; GnRH I; GNRH1; Gonadoliberin I; Gonadorelin; GRH; LHRH; LNRH; Luliberin I; Progonadoliberin 1; Progonadoliberin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6708