Learn More
Biomatik Corporation Porcine PAI1 (Plasminogen Activator Inhibitor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60722
Encompass_Preferred
Porcine PAI1 (Plasminogen Activator Inhibitor 1) ELISA Kit for detecting PAI1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P79335. Target Synonyms: PAI-1; PAI1; SERPINE1; Nexin; PLANH1; Endothelial plasminogen activator inhibitor; PAISerpin E1; PLANH1; plasminogen activator inhibitor 1; serine; or cysteine proteinase inhibitor; clade E; nexin; plasminogenactivator inhibitor type 1; member 1; serpin peptidase inhibitor; clade E; nexin; plasminogen activator inhibitortype 1; member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6713