Learn More
Biomatik Corporation Human EG-VEGF (Endocrine Gland Derived Vascular Endothelial Growth Factor) ELISA Kit
Supplier:  Biomatik Corporation EKF60751
Encompass_Preferred
Human EG-VEGF (Endocrine Gland Derived Vascular Endothelial Growth Factor) ELISA Kit for detecting EG-VEGF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P58294. Target Synonyms: EG-VEGF; PK1; EGVEGF; PRK1; PROK1; Prokineticin 1; Black Mamba Toxin-Related Protein; Mambakine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6724