Learn More
Biomatik Corporation Human SMOX (Spermine oxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF60770
Encompass_Preferred
Human SMOX (Spermine oxidase) ELISA Kit for detecting SMOX in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NWM0. Target Synonyms: SMOX; Spermine oxidasePolyamine oxidase 1; PAO-1; PAOh1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6728