Learn More
Biomatik Corporation Porcine BALP (Bone Alkaline Phosphatase) ELISA Kit
Supplier:  Biomatik Corporation EKF60786
Encompass_Preferred
Porcine BALP (Bone Alkaline Phosphatase) ELISA Kit for detecting BALP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A287BSC3. Target Synonyms: BALP; Bone Alkaline Phosphatase; ALPL; BSAP; HOPS; AP-TNAP; TNSALP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6732