Learn More
Biomatik Corporation Human HS-TNF-α (High sensitive Tumor Necrosis Factor Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60827
Encompass_Preferred
Human HS-TNF-α (High sensitive Tumor Necrosis Factor Alpha) ELISA Kit for detecting TNF-α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.1-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01375. Target Synonyms: TNFα; Tumor Necrosis Factor Alpha; TNF-α; DIF; TNF-alpha; TNFA; TNFSF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6742