Learn More
Biomatik Corporation Human OSCP1 (Protein OSCP1) ELISA Kit
Supplier:  Biomatik Corporation EKF60830
Encompass_Preferred
Human OSCP1 (Protein OSCP1) ELISA Kit for detecting OSCP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8WVF1. Target Synonyms: Protein OSCP1; hOSCP1; Organic solute transport protein 1; Oxidored-nitro domain-containing protein 1; C1orf102; NOR1; OSCP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6744