Learn More
Biomatik Corporation COVID-19 nucleoprotein ELISA Kit
Supplier:  Biomatik Corporation EKF60838
Encompass_Preferred
COVID-19 nucleoprotein ELISA Kit for detecting COVID-19 nucleoprotein in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA. Assay Time: 3h, 30 min. Accession Number: P0DTC9. Target Synonyms: COVID-19 nucleoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6748