Learn More
Biomatik Corporation Mouse Il36b (Interleukin-36 beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60844
Encompass_Preferred
Mouse Il36b (Interleukin-36 beta) ELISA Kit for detecting Il36b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9D6Z6. Target Synonyms: Interleukin-36 beta; Interleukin-1 family member 8 (IL-1F8); Il36b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6751