Learn More
Biomatik Corporation ABHD6 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50008
Encompass_Preferred
ABHD6 Colorimetric Cell-Based ELISA Kit for detecting ABHD6 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9BV23. Gene ID: 57406. Target Synonyms: Monoacylglycerol lipase ABHD6; 2-arachidonoylglycerol hydrolase; Abhydrolase domain-containing protein 6; ABHD6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6761