Learn More
Biomatik Corporation AMPK β1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50028
Encompass_Preferred
AMPK β1 Colorimetric Cell-Based ELISA Kit for detecting AMPK β1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9Y478. Gene ID: 5564. Target Synonyms: 5'-AMP-activated protein kinase subunit beta-1; AMPK subunit beta-1; AMPKb; PRKAB1; AMPK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6777