Learn More
Biomatik Corporation Bcl-6 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50058
Encompass_Preferred
Bcl-6 Colorimetric Cell-Based ELISA Kit for detecting Bcl-6 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41182. Gene ID: 604. Target Synonyms: B-cell CLL/lymphoma 6; B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; Bcl-5; Bcl5; Bcl6; LAZ-3 protein; LAZ3; Zinc finger protein 51; ZNF51. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6802