Learn More
Biomatik Corporation Dab1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50169
Encompass_Preferred
Dab1 Colorimetric Cell-Based ELISA Kit for detecting Dab1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75553. Gene ID: 1600. Target Synonyms: Disabled homolog 1; DAB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6892