Learn More
Biomatik Corporation DAPP1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50171
Encompass_Preferred
DAPP1 Colorimetric Cell-Based ELISA Kit for detecting DAPP1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UN19. Gene ID: 27071. Target Synonyms: 3' phosphoinositide-interacting SH2 domain containing protein; B lymphocyte adapter protein BAM32; Bam32; Dual adaptor for phosphotyrosine and 3-phosphoinositides 1; PHISH; dual adapter for phosphotyrosine and 3-phosphoinositides. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6894