Learn More
Biomatik Corporation Human N-acetyl-β-D-glucosaminidase, NAG ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34722
Encompass_Preferred
The Human N-acetyl-β-D-glucosaminidase, NAG ELISA Kit allows for quantitative determination of N-acetyl-β-D-glucosaminidase, NAG in serum, plasma, urine and tissue homogenates. Species Reactivity: Human. Detection Range: 5 mIU/mL-2000 mIU/mL. Sensitivity: 2 mIU/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07686. Restriction: For Research Use Only.
Catalog No. 50-149-8096