Learn More
Biomatik Corporation ETS1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50214
Encompass_Preferred
ETS1 Colorimetric Cell-Based ELISA Kit for detecting ETS1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14921. Gene ID: 2113. Target Synonyms: C-ets-1 protein; ETS-1; ETS1; p54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6933