Learn More
Biomatik Corporation Human NAD (P)H Dehydrogenase, Quinone 1 (NQO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34725
Encompass_Preferred
The Human NAD (P) H Dehydrogenase, Quinone 1 (NQO1) ELISA Kit allows for quantitative determination of NAD (P) H dehydrogenase, quinone 1 in serum, plasma, tissue homogenates and cell lysates. Species Reactivity: Human. Detection Range: 39 pg/mL-2500 pg/mL. Sensitivity: 9.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15559. Restriction: For Research Use Only.
Catalog No. 50-149-8098