Learn More
Biomatik Corporation hCG β Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50270
Encompass_Preferred
hCG β Colorimetric Cell-Based ELISA Kit for detecting hCG β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01233. Gene ID: 1082; 93659; 94027; 94115. Target Synonyms: Choriogonadotropin subunit beta; CG-beta; Chorionic gonadotrophin chain beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6982