Learn More
Biomatik Corporation HER3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50282
Encompass_Preferred
HER3 Colorimetric Cell-Based ELISA Kit for detecting HER3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21860. Gene ID: 2065. Target Synonyms: c-erbB3; ERB3; ERBB3; HER3; kinase ErbB3; Receptor protein-tyrosine kinase erbB-3 precursor; Tyrosine kinase-type cell surface receptor HER3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6993