Learn More
Biomatik Corporation Histone H4 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50288
Encompass_Preferred
Histone H4 Colorimetric Cell-Based ELISA Kit for detecting Histone H4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P62805. Gene ID: 121504; 554313; 8294; 8359; 8360; 8361; 8362; 8363; 8364; 8365; 8366; 8367; 8368; 8370. Target Synonyms: H4F2; HIST; HIST1H4A; Histone H4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6998